Learn More
Medchemexpress LLC (-)-talarozole | 201410-67-5 | MFCD17166995 | 98.1% | 377.51 g/mol | C21H23N5S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14802D10MG
Encompass_Preferred
(-)-Talarozole is a research-grade small molecule inhibitor of retinoic acid metabolism (RAMBA) used to modulate retinoic acid signaling in biochemical and cellular studies. The compound is the (-)-enantiomer with molecular formula C21H23N5S and molecular weight 377.51 g/mol, and is supplied at high purity for reproducible experimental dosing.
  • Potent inhibitor of retinoic acid metabolism
  • Modulates RAR/RXR signaling and autophagy-related pathways
  • Suitable for in vitro biochemical and cell-based studies
  • Defined molecular formula and molecular weight for accurate dosing
  • High purity appropriate for analytical and assay use
Catalog No. 50-002-87462