Learn More
Medchemexpress LLC Talazoparib tosylate | 1373431-65-2 | 507274 | 99.9% | 552.55 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10841310MM/1ML
Encompass_Preferred
Talazoparib tosylate (BMN 673ts) is a novel, potent, and orally available PARP1/2 inhibitor with an IC50 of 0.57 nM for PARP1. It is for research use only and has not been fully validated for medical applications.
  • Potent PARP1/2 inhibitor
  • Orally available
  • Suitable for research applications
Catalog No. 50-003-47181