Learn More
Medchemexpress LLC HY-131003 5mg Medchemexpress, Taletrectinib CAS:1505515-69-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1310035MG
Encompass_Preferred
Medchemexpress, HY-131003 5mg Taletrectinib CAS:1505515-69-4 Taletrectinib (DS-6051b) is a potent, orally active, and new-generation selective ROS1/NTRK inhibitor. Taletrectinib potently inhibits recombinant ROS1, NTRK1, and NTRK3 with IC50s of 0.207, 0.622, 2.28, and 0.98 nM, respectively. Taletrectinib also inhibits ROS1 G2032R and other Crizotinib-resistant ROS1 mutants[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0245