Learn More
TARGETMOL CHEMICALS INC TALIBEGRON HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T235575MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Talibegron hydrochloride (ZD2079 hydrochloride) is an (beta)3 adrenergic receptor agonist with a pD2 of 3.72 on phenylephrine-preconstricted rat mesenteric artery. Relaxes rat mesenteric artery and isolated aorta in vitro. Inhibits ob gene expression and circulating leptin levels in lean mice in vivo. purity: 99%
Catalog No. 50-258-0858