Learn More
Medchemexpress LLC Talipexole dihydrochloride | 36085-73-1 | 99.80% | C10H17Cl2N3S | 5 MG
Supplier: Medchemexpress LLC HYA00085MG
Talipexole dihydrochloride is a dopamine D2 receptor agonist, an α2-adrenoceptor agonist, and a 5-HT3 receptor antagonist. This compound displays anti-Parkinsonian activity and is a white to off-white solid.
- Targets dopamine D2 receptors
- Acts as an α2-adrenoceptor agonist
- Functions as a 5-HT3 receptor antagonist
- Exhibits anti-Parkinsonian activity
- Soluble in DMSO (25 mg/mL)
- Involved in GPCR/G protein and neuronal signaling pathways
- For research use only