Learn More
Medchemexpress LLC Talipexole dihydrochloride | 36085-73-1 | MFCD00274573 | 99.8% | C10H17Cl2N3S | 100 MG
SDP
Supplier:  Medchemexpress LLC HYA0008100MG
Encompass_Preferred
Talipexole dihydrochloride is a potent dopamine D2 receptor agonist, alpha2-adrenoceptor agonist, and 5-HT3 receptor antagonist. It exhibits significant anti-Parkinsonian activity and is exclusively intended for research applications.
  • Acts as a dopamine D2 receptor agonist
  • Functions as an alpha2-adrenoceptor agonist
  • Serves as a 5-HT3 receptor antagonist
  • Exhibits anti-Parkinsonian activity
  • Suitable for in vitro and in vivo research
  • D2 receptor IC50 value of 25 nM
  • Demonstrated effects on blood pressure and heart rate in feline models
  • Facilitates vagally mediated reflex bradycardia in canine models
Catalog No. 50-004-32452