Learn More
Foxx Life Sciences Borosil® Beaker Griffin Tall Form with Spout, 50mL, 40/CS
Supplier:  Foxx Life Sciences 1060012
Encompass_Preferred
Borosil® Tall-Form Griffin Beakers are designed with pouring spouts for precise pouring. Uniform wall thickness & stress-free glass make them ideal for heating or cooling. High chemical resistance means Borosil® Beakers can be used in nearly any application. Avail. in sizes from 50mL to 1000mL. Borosil® Beakers comply with ISO 3819 & DIN 12331. 500mL is not covered in ISO or DIN standards.In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Low-Expansion Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0340