Learn More
Electron Microscopy Sciences Tall Locking 6 Drawer Cart with Beige Drawers
SDP
Supplier:  Electron Microscopy Sciences 6438697
Encompass_Preferred
White Polyethylene Tall Locking 6 Drawer Cart has a small foot print at 2 square feet, full extension locking drawers and heavy duty industrial casters with brakes. All drawers lock at once and two keys are included. This cart features four 3 inch deep drawers and two 9 inch deep drawers. The top is a 3 inch deep tray with a built in handle for easy maneuvering.
Color: Beige Drawers
Catalog No. 50-192-7335