Learn More
TARGETMOL CHEMICALS INC TALMAPIMOD 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T128715MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Talmapimod (SCIO-469) is an selective orally active and ATP-competitive p38(alpha) inhibitor with IC50 of 9 nM and 90 nM for p38(alpha) and p38(beta) respectively. Talmapimod exhibits at least 2000-fold selectivity over a panel of 20 kinases including other MAPKs. purity: 100%
Catalog No. 50-257-7407