Learn More
Sigma Aldrich Fine Chemicals Biosciences TALONR SuperflowTM Cytiva
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE28957502
Encompass
Talon(R) Superflow is a cobalt-based IMAC medium offering different selectivity compared to nickel-charged media.
Catalog No. 50-290-7190