Learn More
Medchemexpress LLC Taltirelin acetate | 1549593-23-8 | 99.4% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0596A100MG
Encompass_Preferred
Taltirelin acetate is an acetate form of Taltirelin (TA-0910), an orally effective analogue of thyrotropin releasing hormone (TRH). It acts as a TRH receptor superagonist and can cross the blood-brain barrier. Taltirelin acetate stimulates an increase in cytosolic Ca2+ concentration and exhibits neuroprotective effects in cellular and animal models of Parkinson's disease, also alleviating fatigue-like behavior in mouse models of cancer-related fatigue.
  • Orally effective analogue of thyrotropin releasing hormone (TRH)
  • TRH receptor superagonist
  • Crosses the blood-brain barrier
  • Stimulates increase in cytosolic Ca2+ concentration
  • Neuroprotective effects in Parkinson's disease models
  • Alleviates fatigue-like behavior in cancer-related fatigue models
  • Increases cell viability and reduces apoptosis in SH-SY5Y cells
Catalog No. 50-004-09154