Learn More
Medchemexpress LLC Taltirelin acetate (TA-0910 acetate) | 1549593-23-8 | 99.4% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0596A50MG
Encompass_Preferred
Taltirelin acetate (TA-0910 acetate) is an acetate form of Taltirelin (TA-0910), an orally effective analogue of thyrotropin releasing hormone (TRH) and a TRH receptor (TRH-R) superagonist. This compound can cross the blood-brain barrier and stimulates an increase in cytosolic Ca2+ concentration.
  • Orally effective analogue of thyrotropin releasing hormone (TRH)
  • TRH receptor (TRH-R) superagonist
  • Can cross the blood-brain barrier
  • Stimulates an increase in cytosolic Ca2+ concentration (Ca2+ release)
  • Increases cell viability and reduces apoptosis in SH-SY5Y cells and primary rat mesencephalic neurons
  • Demonstrates neuroprotective effects in cellular and animal models of Parkinson's disease
  • Alleviates fatigue-like behavior in mouse models of cancer-related fatigue
Catalog No. 50-004-07654