Learn More
Medchemexpress LLC Taltobulin hydrochloride | 228266-40-8 | 99.3% | 510.11 g/mol | C27H44ClN3O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15584B10MG
Encompass_Preferred
Taltobulin hydrochloride is the hydrochloride salt of taltobulin (HTI-286), a synthetic analogue of the tripeptide hemiasterlin that functions as a potent tubulin (microtubule) inhibitor for preclinical cancer research and ADC payload development.
  • Potent tubulin inhibitor that disrupts microtubule organization.
  • Exhibits low-nanomolar in vitro activity (median IC50 ~1.7 nM).
  • Demonstrated in vivo activity in mouse xenograft models (examples: 1.6 mg/kg i.v., 3 mg/kg p.o.).
  • Supplied as a white to off-white solid for research use.
  • Storage: solid at 4°C, sealed and protected from moisture; solutions stored at -80°C or -20°C for limited durations.
  • High reported purity (~99.3%) suitable for biological studies.
Catalog No. 50-002-04492