Learn More
Medchemexpress LLC Taltobulin hydrochloride | 228266-40-8 | 99.3% | 510.11 g/mol | C27H44ClN3O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15584B50MG
Encompass_Preferred
Taltobulin hydrochloride is a synthetic analogue of the tripeptide hemiasterlin and a potent antimicrotubule agent used in research. It inhibits tubulin polymerization, disrupts microtubule organization, and induces mitotic arrest and apoptosis.
  • Potent tubulin inhibitor for research applications
  • High purity (typically 99.34%)
  • Supplied as a white to off-white solid
  • Available in multiple pack sizes, including 50 mg
  • Used in vitro and in vivo to assess antimitotic activity
Catalog No. 50-002-04312