Learn More
Medchemexpress LLC Taltobulin trifluoroacetate | 228266-41-9 | 99.1% | 587.67 | C29H44F3N3O6 | 10 MG

Supplier: Medchemexpress LLC HY15584A10MG
Taltobulin trifluoroacetate is a potent antimicrotubule research compound that inhibits tubulin polymerization, induces mitotic arrest, and can overcome P-glycoprotein-mediated resistance. It is provided as a solid and as a DMSO solution with defined storage and solubility characteristics for laboratory research use only.
- Potent tubulin inhibitor that induces mitotic arrest.
- Circumvents P-glycoprotein-mediated resistance in cell models.
- Available as a solid (white to off-white) and as a 10 mM solution in DMSO.
- High purity provided (99.1%).
- Molecular weight 587.67, chemical formula C29H44F3N3O6.
- Soluble in DMSO (≥ 39 mg/mL) for stock preparation.
- Storage: solid at 4°C (sealed), solution at -80°C for up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.