Learn More
Medchemexpress LLC Tamarixetin (4'-O-methyl quercetin) | 603-61-2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN118110MG
Encompass_Preferred
Tamarixetin (4'-O-methyl quercetin) is a natural flavonoid research reagent that inhibits caseinolytic protease P (ClpP) and modulates signaling pathways involved in inflammation, apoptosis, and cardiovascular remodeling. It is used as a biochemical probe in infection, cancer, and inflammation studies, and is soluble in DMSO with recommended storage protected from light.
Catalog No. 50-002-96081