Learn More
Selleck Chemical LLC Tamarixetin E2502-1MG
SDP
Supplier:  Selleck Chemical LLC E25021MG
Encompass_Preferred
Tamarixetin (4 -O-Methyl Quercetin) is a natural flavonoid derivative of quercetin and has superior anti-inflammatory properties in bacterial sepsis by increasing the population of IL-10-secreting immune cells
Catalog No. 50-000-40478