Learn More
Indofine Chemical Tamarixetin With Hplc, 10 Mg
SDP
Supplier:  Indofine Chemical 021140S 10 MG
Encompass_Preferred
TAMARIXETIN with HPLC 10 MG, Flavone, Quercetin-4'-methylether; 4'-Methoxy-3,5,7,3'-tetrahydroflavone, C16H12O7, MW: 316.28, 99% by HPLC, 603-61-2, MFCD00017308
Catalog No. 50-909-214