Learn More
US Plastic Corporation TAMCO TRAY 18IN DIA X 8IN H
SDP
Supplier:  US Plastic Corporation 15772
Encompass
Welded constructionCapacity: 8.5 gallonsWall thickness: 3/16" bottom & 1/8" sidesRigid, sturdy constructionCorrosion resistantDimensions are inside dimensionsMaximum temperature: 140°FMaterials meet FDA standardsTrays & covers sold separately
Catalog No. NC2390296