Learn More
Selleck Chemical LLC Tamibarotene 50mg 94497-51-5
SDP
Supplier:  Selleck Chemical LLC S426050MG
Encompass_Preferred
Tamibarotene is a synthetic retinoic acid receptor (RAR) agonist with high specificity for RAR? and RAR? over RAR?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3814