Learn More
Selleck Chemical LLC Taminadenant E4632-100MG
SDP
Supplier:  Selleck Chemical LLC E4632100MG
Encompass_Preferred
Taminadenant (NIR178 PBF509) is a non-xanthine and non-furan antagonist of adenosine A2A receptor (A2AR) It can antagonize A2AR agonist-mediated cAMP accumulation and impedance responses with Kb values of 72 8 nM and 8 2 nM respectively It exhibits potential utility for Parkinson s disease (PD) treatment
Catalog No. 50-000-39415