Learn More
Medchemexpress LLC Tamnorzatinib 10mM 1mL | 1646839-59-9 | 1 ML

Supplier: Medchemexpress LLC HY11435810MM 1ML
Tamnorzatinib is a potent, selective inhibitor of the Axl and Mer receptor tyrosine kinases, supplied as a 10 mM solution in DMSO for research applications. It is used in biochemical and cellular assays to study Axl/Mer signaling and related oncology pathways.
- Potent Axl and Mer kinase inhibition with low-nanomolar activity.
- Supplied as a 10 mM solution in DMSO for convenience in assays.
- High purity, suitable for research applications.
- Chemical formula C32H26N4O6 and molecular weight 562.57 g/mol.
- CAS number 1646839-59-9 for unambiguous identification.
- Provided as a 1 mL aliquot; store per recommended conditions to preserve stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.