Learn More
Cayman Chemical Tamoxifen, 10540-29-1, 5 g
SDP
Supplier:  Cayman Chemical 132585
Encompass_Preferred
A selective estrogen receptor (ER) modulator; an ER antagonist in breast tissue, effective in treating early breast cancer; an ER agonist in bone and blood vessels and a partial ER agonist in uterine tissues
Catalog No. NC0601169