Learn More
Sigma Aldrich Fine Chemicals Biosciences Tamoxifen, 10540-29-1, MFCD00010454, 5 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T56485G
Encompass
Linear Formula: C6H5C(C2H5)=C(C6H5)C6H4OCH2CH2N(CH3)2, Molecular Weight: 371.51, Synonym: (Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene, trans-2-[4-(1,2-Diphenyl-1-butenyl)phenoxy]-N,N-dimethylethylamine, Assay ≥99%. Tamoxifen has been used to facilitate the recombination of ect2flox allele in mouse organs. It has also been used to study its effect on lipopolysaccharide (LPS)-induced microglial activation.
Catalog No. NC0827840