Learn More
Enzo Life Sciences Tamoxifen. citrate (1g). CAS: 54965-24-1
SDP
Supplier:  Enzo Life Sciences ALX550095G001
Encompass_Preferred
Antiestrogen with anti-cancer activity. Selective protein kinase C (PKC) inhibitor and induces PKC epsilon translocation. Induces apoptosis in estrogen receptor-negative human cancer cell lines. Alternative name: (Z)-2-[4-(1,2-Diphenyl-1-butenyl)phenoxy]-N,N-dimethyl-ethanamine . citrate. Purity: ≥99% (Assay). Appearance: White to off-white powder. Solubility: Soluble in methanol or ethanol; very slightly soluble in water or acetone. Long Term Storage: +4°C.
Catalog No. 50-200-9481