Learn More
Selleck Chemical LLC Tamoxifen Citrate 10mM/1mL 54965-24-1 ICI 46474 Citrate
SDP
Supplier:  Selleck Chemical LLC S197210MM/1ML
Encompass_Preferred
Tamoxifen Citrate (ICI 46474) is a selective estrogen receptor modulator (SERM). Tamoxifen Citrate is also a potent Hsp90 activator and enhances the Hsp90 molecular chaperone ATPase activity. Tamoxifen induces apoptosis and autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1823