Learn More
Abcam Tamo x ifen citrate, Estrogen receptor antagonist and partial agonist, 100MG
Supplier:  Abcam AB120656100MG
Encompass_Preferred
MW 563.65 g/mol, Purity >99%. Competitive estrogen receptor antagonist and partial agonist. Inhibits estrogen dependent gene transcription and tumor growth. Additionally decreases cardiac infarction and inhibits Kir2.1, Kir2.2 and Kir2.3 potassium channels (IC₅₀ values are 0.93, 0.87 and 0.31 μM, respectively).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7383