Learn More
Medchemexpress LLC Tamoxifen-d3 hydrochloride | 00-00-0 | 99.0% | 410.99 | C26H27D3ClNO | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13757S5MG
Encompass_Preferred
Tamoxifen-d3 hydrochloride is a deuterium-labeled form of tamoxifen provided as a high-purity hydrochloride salt for use as a tracer or internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS. It is supplied as a white to off-white solid with specified solubility and storage recommendations to preserve isotopic integrity.
  • Deuterium-labeled tracer for quantitative analysis
  • High purity minimizes analytical interference
  • Supplied as a stable hydrochloride solid for handling convenience
  • Soluble in DMSO; ultrasonic warming recommended for dissolution
  • Recommended cold, sealed storage to maintain stability
Catalog No. 50-002-22087