Learn More
Medchemexpress LLC Tamoxifen-d3 hydrochloride | 00-00-0 | 99.0% | C26H27D3ClNO | 10MG
SDP
Supplier:  Medchemexpress LLC HY13757S10MG
Encompass_Preferred
Tamoxifen-d3 hydrochloride is a deuterium-labeled analogue of tamoxifen supplied as a white to off-white solid. It is used as an internal standard and tracer in mass spectrometry, pharmacokinetic studies, and analytical method development, providing a distinct mass shift from the unlabeled compound for accurate quantitation and identification.
  • Deuterium-labeled for mass-spectrometry differentiation.
  • High purity suitable for analytical standards and tracer studies.
  • Solid form simplifies handling and storage.
  • Stable under recommended storage conditions for reproducible results.
  • Suitable for pharmacokinetic and bioanalytical method development.
Catalog No. 50-002-59655