Learn More
Cayman Chemical TamoxIfen N-oxIde 10mg
SDP
Supplier:  Cayman Chemical 1864210 MG
Encompass_Preferred
A metabolite of tamoxifen that is produced by the action of flavin-containing monooxygenases
Catalog No. 50-297-9958