Learn More
Medchemexpress LLC Tamoxifen N-oxide | 75504-34-6 | MFCD00869497 | 97.8% | 387.51 g·mol⁻¹ | C26H29NO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1330105MG
Encompass_Preferred
Tamoxifen N-oxide is a research-grade metabolite of tamoxifen used as a reference standard in metabolic, pharmacokinetic, and analytical studies. It is supplied as a purified solid suitable for analytical assays and includes documentation for quality control and safe handling.
  • High-purity reference compound for analytical and metabolic studies.
  • Useful as a metabolite standard in pharmacokinetic and drug metabolism research.
  • Suitable for use in LC-MS and other analytical assays.
  • Supplied in small-mass vials appropriate for research dosing and method development.
  • Documentation available: datasheet, certificate of analysis, and safety data sheet.
  • Molecular formula C26H29NO2; molecular weight 387.51 g·mol⁻¹.
Catalog No. 50-002-62271