Learn More
Medchemexpress LLC Tamra-PEG2-maleimide | 2304558-24-3 | 95.0% | 640.68 g/mol | C35H36N4O8 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW1909321MG
Encompass_Preferred
TAMRA-PEG2-Maleimide is a TAMRA-derived fluorescent dye bearing a PEG2 spacer and a maleimide reactive group for selective conjugation to thiol (-SH) groups. Supplied as a solid, it is used for site-specific labeling of cysteine residues, peptides, and other thiol-containing molecules for fluorescence-based assays and imaging.
  • Maleimide functional group for selective thiol conjugation.
  • TAMRA fluorophore for visible-range fluorescence detection.
  • PEG2 spacer improves solubility and reduces steric hindrance.
  • High purity suitable for bioconjugation (95.0%).
  • Soluble in DMSO at ≥ 100 mg/mL for convenient stock preparation.
  • Store protected from light at recommended temperatures for stability.
Catalog No. 50-002-83140