Learn More
TARGETMOL CHEMICALS INC TAMRA-PEG3-Azide 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T169821MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TAMRA-PEG3-Azide is a TAMRA red fluorescent dye linker with a maximum excitation/emission wavelength of 553/575 nm. Purity 95%
Catalog No. 50-287-9378