Learn More
Medchemexpress LLC Tamra-PEG4-acid | 1909223-02-4 | MFCD31536753 | 95.3% | C36H43N3O10 | 10MG

Supplier: Medchemexpress LLC HY14050910MG
TAMRA-PEG4-acid is a red-fluorescent dye bearing a PEG4 spacer and a terminal carboxylic acid, formulated for covalent labeling of biomolecules and probe synthesis. It offers a hydrophilic linker to reduce steric interference and improve solubility, and is provided at high purity for analytical and imaging workflows.
- Provides red fluorescence with excitation/emission around 553/575 nm.
- Contains a four-unit polyethylene glycol spacer to increase hydrophilicity.
- Terminal carboxylic acid enables standard amide and ester coupling chemistries.
- High purity suitable for labeling, imaging, and analytical applications.
- Soluble in DMSO; ultrasonic treatment and formulation protocols improve aqueous compatibility.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.