Learn More
Cayman Chemical TAMRAAmyloId128 PeptId 100ug
SDP
Supplier:  Cayman Chemical 27110100 UG
Encompass_Preferred
A fluorescently labeled amyloid-β peptide; ex/em = 543/572 nm
Catalog No. 50-294-0791