Learn More
Leica Biosystems Snowcoat Microscope Slides, Tan, 25 x 75 mm (1 x 3 in), 1,440/pack
Supplier:  Leica Biosystems 3800363
Encompass
Snowcoat microscope slides are 25 x 75 mm glass slides with a colored snowcoat marking area for clear labeling and color-coded identification. They offer consistent thickness for reliable specimen mounting and are suitable for histology and general microscopy workflows.
  • Colored marking area for easy labeling and color coding.
  • Compatible with xylene-resistant pens and pencils for durable markings.
  • Standard 25 x 75 mm size fits common slide holders and printers.
  • Consistent thickness for reliable specimen mounting and imaging.
  • Bulk packaging supports high-throughput histology workflows.
Catalog No. NC3237841