Learn More
Medchemexpress LLC Tandospirone | 87760-53-0 | MFCD00904852 | 99.4% | 383.49 g/mol | C21H29N5O2 | 5 MG

Supplier: Medchemexpress LLC HY145585MG
Tandospirone is a selective 5-HT1A receptor partial agonist used in preclinical pharmacology and neuroscience research. It is supplied with characterized purity and recommended storage conditions for laboratory use only; not for human consumption.
- Selective 5-HT1A receptor partial agonist.
- High purity: 99.4%.
- Cas number: 87760-53-0.
- Molecular formula: C21H29N5O2.
- Molecular weight: 383.49 g/mol.
- Available formats: solid powder and in solvent.
- Typical pack size: 5 MG.
- Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 2 years) or -20°C (up to 1 year).
- Intended for research use only; not for clinical or human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.