Learn More
ATCC Shouchella clausii
Supplier:  ATCC 700160
Encompass
Shouchella clausii strain DSM 8716 is a whole-genome sequenced bacterial type strain that was isolated from garden soil in Denmark.
Catalog No. 50-237-9586