Learn More
ATCC Alicyclobacillus acidocaldarius
Supplier:  ATCC 43033
Encompass
Alicyclobacillus acidocaldarius strain NRS 1664 [GD1B] was isolated in 1971 from a hot spring in Yellowstone National Park. This thermo-acidophilic bacterium has applications in biotechnology.
Catalog No. 50-238-0484