Learn More
Medchemexpress LLC Tanshindiol C | 97465-71-9 | 98.5% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1229365MG
Encompass_Preferred
Tanshindiol C is a small-molecule research compound reported to inhibit EZH2 (histone methyltransferase). It is supplied as a high-purity analytical reagent for in vitro and preclinical research.
  • High purity suitable for analytical and biological assays
  • Defined molecular weight 312.32 for accurate calculations
  • Molecular formula C18H16O5 consistent with characterization data
  • Available in small quantities for screening and dose-response studies
  • Provided as a solid for stable storage and handling
Catalog No. 50-002-30518