Learn More
BERTECH KELEX 1/2 In. Wide x 36 Yards Long, 5 Mil Thick Kapton Tape
SDP
Supplier:  BERTECH KELEX KPT51/2
Encompass_Preferred
Kapton Tapes are made from 5 Mil thick Kapton HN film with silicone adhesive. They have been used in applications at temperatures as low as -73°C (-99°F) and as high as 260°C (500°F). DuPont™ Kapton Polyimide Films have set the industry standard for over 45 years in high performance, reliability and durability, with a unique combination of electrical, thermal, chemical and mechanical properties that withstand extreme temperatures, vibration, and other demanding environments. Kapton Tapes have a high dielectric strength. The silicon adhesive protection leaves no residue. Also available in 1 mil and 2 mil thickness.Applications: 3D Printing, Aerospace Assembly, Automotive Facilities, Electronic Assembly, Fiber Optics Manufacturing and Solar Panels.
Catalog No. NC2110274