Learn More
Med Vet International Waterproof Medical Tape 3M Blenderm Transparent 1 inch X 5 Yard Plastic Nonsterile, 1525-1, 120/Case
Supplier:  Med Vet International 5758CS
Encompass_Preferred
The 3M Blenderm surgical tape is waterproof to keep out moisture and contaminants It's conformable to provide coverage in hard-to-tape areas This tape is useful for securing devices and dressings Its clear design provides transparency The medical tape is hypoallergenic to be gentle on sensitive skin 1 inches x 5 yards
Catalog No. 50-283-1640