Learn More
Paul N Gardner Company TAPE ADHESION TEST KIT
SDP
Supplier:  Paul N Gardner Company PA2000
Encompass
Paint Adhesion Test Kit includes blade handle with guide (blade not included), hex wrench for changing blades, flaking brush, illuminated magnifier with batteries, tape, copy of ASTM Test Method D-3359 for use as an instruction guide, and a sturdy plastic carrying case with handle.
Catalog No. NC9854940