Learn More
Med Vet International Medical Tape 3M Durapore White 1 inch X 1-1/2 Yard Silk-Like Cloth Nonsterile, 1538S-1, Each
Supplier:  Med Vet International 6020EA
Encompass_Preferred
3M Durapore surgical tape is well-suited for securing high-strength applications The nonstretch silk-like cloth retains its shape for reliable performance This hypoallergenic 3M Durapore medical tape is safe for sensitive skin The material conforms to the body letting you achieve a personalized fit Bi-directional tear for quick use The white tape can be torn while wearing gloves because it won't stick to gloves 1 inch x 1-1/2 yards
Catalog No. 50-283-0272