Learn More
Grainger Tape Brown 50 ft L 4 in W
Supplier:  Grainger 14A593
Encompass_Preferred
The Brady Continuous Label Roll is a 4 inch wide white polyester label roll that is ideal for wire and cable marking, panel labeling, equipment ID, and general purpose identification. This label roll features a permanent acrylic adhesive and comes in a length of 250 feet. The material is resistant to chemicals, abrasion, and temperatures up to 250°F. It can be used with the Brady label printers for easy custom printed labels on demand.
Catalog No. 50-275-3624