Learn More
Grainger Tape Brown 50 ft L 6 in W
Supplier:  Grainger 14A597
Encompass_Preferred
The Brady Continuous Label Roll is a 6 inch wide white polyester label roll that is ideal for wire and cable marking, panel labeling, equipment ID, and general purpose identification. This label roll features a permanent acrylic adhesive and comes in a length of 250 feet. It is designed for use with the Brady label printers and can withstand temperatures from -40°F to 250°F. The polyester material and acrylic adhesive ensure the labels will adhere well and resist chemicals, abrasion, and moisture. With high-contrast black ink, these customizable labels will create durable and legible identification.
Catalog No. 50-275-3625