Learn More
Grainger Tape Cartridge Phosphorescent 50 ft L
Supplier:  Grainger 14A413
Encompass_Preferred
The Brady Label Tape Cartridge is a self-laminating vinyl label tape that creates durable, fade-resistant labels for wire and cable marking, panel and equipment identification, safety and facility identification, and more. It features a 1" x 10' length of white tape with black text, is compatible with the Brady TLS2200 thermal transfer printer, and creates labels that resist chemicals, abrasion, moisture, and extreme temperatures. The cartridge contains no adhesives and allows for quick, clean label application. Overall, this product provides a convenient and reliable labeling solution for a variety of industrial applications.
Catalog No. 50-275-3521