Learn More
Med Vet International Medfix Retention Dressing Tapes, 2in x 11 yds. 1/bx
Supplier:  Med Vet International MSC4002
Encompass_Preferred
For flexibility and customized sizing. Low sensitivity adhesive, gentle for the patient. Medfix™ has a printed s-c release liner. Water resistant. Indications: to secure primary dressings and to secure gastrostomy tubes and otherfeeding tubes. Contraindicated as a primary dressing. May be left in place for up to 7 days. Dressing change frequency will depend on amount of drainage.
Catalog No. 50-252-2257