Learn More
Med Vet International 3M Fiberglass Casting Tape 4" X 4 Yds
Supplier:  Med Vet International 3MFG4
Encompass_Preferred
A lightweight, strong and durable casting tape that combines the benefits of fiberglass casting tape with the handling ease of plaster. Made from water-activated resin which allows easier, tack-free application. No choice of color.
Catalog No. 50-283-0171