Learn More
Grainger Tape Gold 50 ft L 8 in W
Supplier:  Grainger 14A594
Encompass_Preferred
The Brady Continuous Label Roll is an 8 inch wide white polyester label roll that prints using thermal transfer printers. It has a permanent acrylic adhesive and is designed for wire and cable marking, panel labeling, equipment ID, and general purpose identification. The roll is 8 inches wide by 300 feet long. It has a service temperature range of -40°F to 302°F and is suitable for indoor and outdoor use. The labels have a matte finish and feature a strong permanent adhesive that adheres well to a variety of surfaces including curved, textured, and powder coated. It can be used with the Brady TLS 2200, IDXPERT, BMP21-PLUS, and BMP21 printers.
Catalog No. 50-275-1720